Would you like to log in again?
Ward's® Prepared Nematode Growth Medium
Supplier: MASTERDATA QL HOLD 2
Ward's®
WARD470180-715 WACP470180-736
470180-736EA 14.55 USD InStock
470180-736 470180-715
Ward's® Prepared Nematode Growth Medium
Educational Materials Biology Educational Materials Microbiology Learning Activities
Designed for cultivation of Ceanorhabditis elegans.
  • Sterile - no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Prepared nematode growth medium, available as sterile agar plates and agar bottles. Great for aseptic culturing of Caenorhabditis elegans.
Ordering information: During checkout, please specify a delivery date for your specimens to arrive.
Delivery information: Prepared plates will ship via overnight or 2nd day delivery, prepared bottles do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of shipping.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty