Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BACTIDENT™ Rapid Tests: Indole and Oxidase Reagents

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1.00181.0002 1.11350.0001
470180-774EA 54.9 USD InStock
470180-774 470194-786
BACTIDENT™ Rapid Tests: Indole and Oxidase Reagents
Environmental Test Kits
Indole Reagent
Indole Reagent for the Indole reaction of Enterobacteria. 30 ml bottle.

Oxidase Test Strips
Test Strips to detect production of cytochrome oxidase in microorganisms. Package of 50 strips.

Reagents ship at ambient temperature. Store at +2°C to +8°C.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty