Would you like to log in again?
Marchantia, Mature Sporophyte Slide
Supplier: Avantor
WARD470181-120
470181-120EA 6.95 USD InStock
470181-120
Marchantia, Mature Sporophyte Slide
Slides Prepared Slides
Life Cycle Phase of Liverwort
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Liverwort. Shows foot, seta, sporangium, spores, and elaters of sporangia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty