Would you like to log in again?
Zebrafish Slides (Danio rerio)
Supplier: Avantor
WARD470181-700 WARD470181-708 WARD470181-706 WARD470181-710
470181-708EA 13.49 USD InStock
470181-708 470181-706 470181-700 470181-710
Zebrafish Slides (Danio rerio)
Slides Prepared Slides
Zebrafish are model organisms for vertebrate and genetic research.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Zebrafish (Danio rerio) produce many offspring and have a short generation time for long term studies
We offer individual zebrafish slides of:
▪ Zebrafish: The entire fish is visible to the naked eye, but when viewed under a microscope, students can see the wide diversity of cell types found in one animal.
▪ Zebrafish, Hatchling : Typical hatchling, entire specimen.
▪ Zebrafish, Cleavage Stage: Illustrates cleavage stage.
▪ Zebrafish, Embryo: Entire embryo with yolk sac.
Embryos are great models to study vertebrate development because they are mostly transparent and developmental stages are easily visible.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty