Would you like to log in again?
Ward's® Frog Anatomy Slides
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
WARD470181-742 WARD470181-758 WARD470177-036 WARD470181-754 WARD470007-796 WARD470182-388 WARD470181-740 WARD470182-390 WARD470181-756 WARD470182-392 WARD470177-536 WARD470181-738 WARD470181-752 WARD470181-770
470182-388EA 11.5 USD InStock
470182-388 470182-392 470177-536 470181-738 470181-758 470177-036 470007-796 470182-390 470181-756 470181-754 470181-742 470181-752 470181-740 470181-770
Ward's® Frog Anatomy Slides
Slides Prepared Slides
Microscopically study frog and amphibian features that best enhance your curriculum.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Frogs have helped scientists learn about the nervous and circulatory systems, vertebrate anatomy, and reproductive hormones in historic studies. Frogs have adaptations to help them survive, like mucosal glands to prevent moisture loss and poison glands in the skin to defend against predators. Amphibian blood, unlike mammalian blood, is nucleated and is a useful feature in amphibian and comparative physiology studies.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty