Would you like to log in again?
Sarcocystis Slide
Supplier: Avantor
WARD470181-966
470181-966EA 14.49 USD InStock
470181-966
Sarcocystis Slide
Slides Prepared Slides
Asexual Multiplication within the sarcocyst Occurs
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Striated muscle shows thread-like sarcocystis, historically known as Miescher’s tubules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty