Would you like to log in again?
Taenia pisiformis, Mature/Gravid Proglottids Slide
Supplier: Avantor
WARD470182-112
470182-112EA 19.69 USD InStock
470182-112
Taenia pisiformis, Mature/Gravid Proglottids Slide
Slides Prepared Slides
Endoparasitic Tapeworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Composite of mature and gravid proglottids of Taenia pisiformis. Mature proglottid shows uterus, ovary, testes, sperm duct, and genital pore. Gravid proglottid shows eggs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty