Would you like to log in again?
Taenia pisiformis, Eggs Slide
Supplier: Avantor
WARD470182-114
470182-114EA 9.65 USD InStock
470182-114
Taenia pisiformis, Eggs Slide
Slides Prepared Slides
Endoparasitic Tapeworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Egg of Taenia pisiformis. The so-called “shell” is actually an embryonic membrane, the embryophore. The embryo is within. Unstained.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty