Would you like to log in again?
Esophagus-Stomach Junction, Mammal Slide
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470182-852
470182-852EA 7.6 USD InStock
470182-852
Esophagus-Stomach Junction, Mammal Slide
Slides Prepared Slides
Illustrates a section of the digestive system
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Demonstrates the transition from the stratified squamous epithelium of the esophagus to the simple columnar epithelium of the beginning of the stomach.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty