Would you like to log in again?
Lung Slide
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470182-904
470182-904EA 7.95 USD InStock
470182-904
Lung Slide
Slides Prepared Slides
Lung characteristics illustrated
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal lobule. Shows alveolar sacs and small bronchioles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty