Would you like to log in again?
Kidney, Mammal Slide
Supplier: Avantor
WARD470182-924
470182-924EA 13.5 USD InStock
470182-924
Kidney, Mammal Slide
Slides Prepared Slides
Investigate kidney structure
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. For detailed study of cortex and medulla. Shows renal corpuscle, tubule, and collecting duct.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty