Would you like to log in again?
Barr Bodies Absent (SM) H and E Slide
Supplier: MASTERDATA QL HOLD 2
7938111
470183-142EA 5.59 USD InStock
470183-142
Barr Bodies Absent (SM) H and E Slide
Slides Prepared Slides
Smear of isolated cells from buccal cavity of a normal human male. Barr bodies are absent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty