Would you like to log in again?
Chemistry of Life Slide Set
Supplier: Avantor
WARD470183-408
470183-408KT 97.95 USD InStock
470183-408
Chemistry of Life Slide Set
Slides Prepared Slides
Examples from the Major Categories of Biochemical Composition
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All slides are prepared as sections unless otherwise specified.
Ordering information: Set includes Carbo:hydrates Glycogen, liver; Mucous, small intestine goblet cells; Lipids: Droplet, adipose tissue; Membrane, peripheral nerve myelin (cs); Proteins: Anti-tyrosine hydroxylase, neuron; Anti-neurofilament, spinal cord; Nucleic Acids: Nucleus/ribosomes, neuron (cs); and Chromosomes, blastodisc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty