Would you like to log in again?
Genetically Modified Corn Slide Set
Supplier: Avantor
WARD470183-452
470183-452KT 48.99 USD InStock
470183-452
Genetically Modified Corn Slide Set
Slides Prepared Slides
Connect Molecular Biology Concepts to Real-World Examples
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
This slide set displays both examples of common vectors adapted to introduce modifications into an important food target, as well as the cellular structures of that target.
Ordering information: Set includes Agrobacterium tumefaciens (sm), Bacillus thuringiensis (sm), Crown gall (sect), Zea (Corn) Stem (cs), Genetically Modified Zea Stem (cs), Zea (Corn) Leaf (cs), and Genetically Modified Zea Leaf (cs).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty