Would you like to log in again?
Comparative Plant Anatomy Slide Set
Supplier: Avantor
WARD470183-472
470183-472KT 67 USD InStock
470183-472
Comparative Plant Anatomy Slide Set
Slides Prepared Slides
Illustrate Basic Morphologic Divergence Between the Monocot and Dicot Plant Groups
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Ordering information: Set includes Monocot and dicot roots (cs); Primary and secondary stems (cs); Dicot and monocot stems (cs); Dicot and monocot leaves (cs); Air and water leaves (cs); and Leaf types (cs).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty