Would you like to log in again?
Cell Division Slide Set
Supplier: Avantor
WARD470183-556
470183-556KT 38 USD InStock
470183-556
Cell Division Slide Set
Slides Prepared Slides
Unique Comparison of Plant and Animal Mitosis
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Ordering information: Set includes Plant (onion) mitosis (ls); and Animal (whitefish blastodisc) mitosis (sect).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty