Would you like to log in again?
General Nerve Tissues Slide Set
Supplier: Avantor
WARD470183-592
470183-592EA 344 USD InStock
470183-592
General Nerve Tissues Slide Set
Slides Prepared Slides
Expanded Set Explores Functions and Arrangement of Neurons
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Ordering information: Set includes Motor Neurons: Giant multipolar motor neurons (sm); Motor nerve endings (Whole mount); Nissl bodies (sect); Neurofibrillae (sect); Neuron Types: Pyramidal cells and their processes (sect); Purkinje cells and their processes (sect); Glial Cells: Neuroglia, protoplasmic astrocytes (sect); Medullated Nerves: Medullated nerve fibers, teased peripheral nerves (Whole mount); Medullated nerve (cs/ls); Peripheral nerve, treated with osmic acid (cs); Peripheral nerve (cs); Sensory Neurons: Pacinian corpuscle, pressure (Whole mount/cs); Neuro-muscular spindle, sensory end-organ in muscle (Whole mount); Retina (cs); Cochlea (sect); Olfactory epithelium (sect); Tongue (sect); Cerebrum sensory area (sect); Ganglia: Auerbach’s plexus, parasympathetic (sect); Spinal ganglion (ls); Sympathetic ganglion (ls); Spinal Cord: (5 slides; including 4-level composite); Cerebellum (4 slides); Cerebrum (2 slides); Medulla (2 slides).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty