Would you like to log in again?
Colon, Mammal, Thin Section Slide
Supplier: Avantor
WARD470183-688
470183-688EA 16.5 USD InStock
470183-688
Colon, Mammal, Thin Section Slide
Slides Prepared Slides
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty