Would you like to log in again?
Graduated Cylinders with Plastic Base
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CY137-10 CY137-50 CY137-250 CY137-50 CY137-1000 CY137-500 CY137-100 CY137-25
470349-758EA 20.75 USD InStock
470349-758 470188-955 470121-934 76531-552 470349-806 470188-950 470188-954 470188-952
Graduated Cylinders with Plastic Base
Cylinders Measuring Cylinders
Durable Borosilicate Glass Construction.
  • Graduations permanently fused into glass
  • Hexagonal base and bumper guard add protection for all sizes
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty