Would you like to log in again?
AP Drosophila Crosses- Live
Supplier: VWR
WARD470189-288 WARD470189-284 WARD470189-286
470189-284EA 31.95 USD InStock
470189-284 470189-286 470189-288
AP Drosophila Crosses- Live
Specimens Live Specimens Animals - Living Material
Four to six adults of each sex (including virgin females) have been carefully selected for the parent generation. Larvae will be visible within a week of receipt. Please order 7-10 days in advance.
We fully guarantee the purity and viability of our Drosophila melanogaster cultures. A large, vigorous culture is shipped in a shatterproof plastic vial. To ship Drosophila cultures with living larvae we require 10 days' notice prior to use date. During very cold weather, adult flies may die but eggs and larvae usually survive. Allow cultures to come to room temperature before checking under low magnification for eggs and live larvae. Complete instructions are always included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty