Would you like to log in again?
Enzyme Catalysis Lab Activity
Supplier: Edvotek
282
470189-532KT 179 USD InStock
470189-532
Enzyme Catalysis Lab Activity
Educational Materials Biology Educational Materials AP Biology Learning Activities
In this experiment, students safely analyze the enzymatic decomposition of hydrogen peroxide.
  • Material for 10 Lab Groups
  • Calorimetric Assay Avoids Potential Dangers
  • Estimated Required Lab Time: 30 to 45 Minutes
  • No Corrosive Acid
  • Expensive Burettes and Syringes Not Needed
Students use a calorimetric assay that avoids potential dangers associated with the use of corrosive acid or potassium permanganate or the expense of burettes and syringes. They measure reaction rates and discuss the effects of parameters such as temperature and substrate concentration.
The activity is meant for 10 lab groups.
Ordering information: This lab includes a coupon for perishable items, catalase solution, hydrogen peroxide, a phosphate buffer, assay reagent A, cidification solution, color enhancer and developer concentrate, a plastic tube, and instructions. A visual spectrophotometer, timers, beakers, 10 mL test tubes with racks, 1, 5 and 10 mL pipets with bulbs or pumps, linear graph paper, markers, distilled water, ice, and personal safety equipment are needed but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty