Would you like to log in again?
Live Conocephalum Clump
Supplier: GULF OF MAINE, INC.
CONO3X4
470191-762EA 28.2 USD InStock
470191-762
Live Conocephalum Clump
Specimens Live Specimens Plants - Living Material
Conocephalum is an ideal specimen for illustrating alternation of generations in Bryophytes.
  • Prefers moist soil and temperate regions
  • Excellent for use as a substrate for classroom terraria
Conocephalum sp. The vegetative stage of this bryophyte features dichotomously branched, flat, ribbon-like thallus, like Marchantia. The thallus is differentiated with elongated “root” cells on the ventral surface that anchor the plant.
Ordering information: These products contain living or perishable materials. Please specify a delivery date at order time.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty