Would you like to log in again?
Transformation of Escherichia coli with Green or Blue Fluorescent Protein, Edvotek
Supplier: Edvotek
222 222
470200-176KT 179 USD InStock
470200-176 470193-130
Transformation of Escherichia coli with Green or Blue Fluorescent Protein, Edvotek
Educational Classroom Kits and Activities
Using this kit, students genetically engineer bacteria with genes from a jellyfish.
  • GFP and Mutated BFP Plasmids Included
  • Guided or Open-Inquiry
  • At Room Temperature, Bactobeads Remain Stable for Nine Months
  • Time Required: 90 Minutes Over Two to Three Days
  • Students Transform Bacteria with GFP, BFP, or Both
The Green Fluorescent Protein (GFP), which is responsible for bioluminescence in the jellyfish Aequorea victoria, is used extensively in all areas of science. Many organisms have been transformed with the GFP gene.
It's so useful that scientists have mutated it to produce Blue Fluorescent Protein (BFP). In this simple experiment, students transform bacteria with GFP, BFP, or both.
Ordering information: This kit includes instructions, BactoBeads, plasmid DNA, IPTG, ampicillin, transformation solution, ReadyPour agar, Luria broth, petri dishes, sterile pipettes and loops, email coupon redeemable for perishable components. A water bath, 37°C incubation oven, microwave or hot plate, and long-wave UV lamp are required but not included.
Delivery information: Some components require refrigerator and freezer storage on arrival.
Packaging: Some components require refrigerator & freezer storage on arrival
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty