Would you like to log in again?
Potato Dextrose Agar
Supplier: IPM SCIENTIFIC, INC.
880007
470193-232EA 30.85 USD InStock
470193-232
Potato Dextrose Agar
Media for Microbiology Dehydrated Media for Microbiology
For cultivation of fungi and yeasts.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 .
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty