Would you like to log in again?
Coliform in Water Test Lab Activity
Supplier: MICROLOGY INC
K24WQ
470193-380KT 134.15 USD InStock
470193-380
Coliform in Water Test Lab Activity
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Students apply lab work to real-life issues.
  • Students Collect Local Water Samples and Tests for Coliform Bacteria
  • Test Results Compared to Safe and Unsafe Quantities
  • Sewage Treatment Facilities Test for the Same Bacterial Group
  • Links to Real-World Scenarios
  • Teacher's Manual Included
They identify a bacterial group for which actual sewage treatment facilities test.
The class samples local water for coliform bacteria then compares results to a chart showing safe and unsafe quantities.
Ordering information: This kit includes instruction and material for a class of 24 students including Easygel, a pre-measured, sterilized culture media that requires no mixing or autoclaving.
Delivery information: This kit includes perishable materials. Please specify delivery date.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty