Would you like to log in again?
Salvinia
Supplier: VWR
865800
470194-686EA 10.95 USD InStock
470194-686
Salvinia
Specimens Live Specimens Plants - Living Material
Salvinia rotundifola. This floating water fern has paired oblong leaves that are covered with hairs on both sides. The top part of the leaves are green, the underside purple. Salvinia was originally native to South America and has been introduced in the United States. It is adapted to ponds, pools, and marshes. It is supplied in a 4 oz. jar with care instructions. Note: Not available in Canada.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty