Would you like to log in again?
Ward's® Physiology of the Circulatory System Lab Activity
Supplier: MASTERDATA QL HOLD 2
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470194-706
470194-706EA 35.95 USD InStock
470194-706 470009-226
Ward's® Physiology of the Circulatory System Lab Activity
Educational Materials Biology Educational Materials AP Biology Learning Activities
View Daphnia Heart Rate Easily
  • Teaches technique for measuring blood pressure
  • Comparative physiology featuring Daphnia
  • Specifically designed viewing chambers for viewing Daphnia
  • Time Required: 75 minutes
Students learn how to measure blood pressure, pulse, and heart rate, and assess the effects that varying physiological conditions have on their rates, in this practical, comprehensive lab activity. For comparison, students also study the heart rate of Daphnia, varying the temperature and viewing the subsequent changes that take place in the ectothermic creature. Ward's has included the components of a viewing chamber that students make themselves for the easiest viewing of the Daphnia specimens.
Materials required, not included: sphygmomanometer (blood pressure cuff)
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty