Would you like to log in again?
Prepared Eosin Methylene Blue Agar (EMB) Media
Supplier: IPM SCIENTIFIC, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
880960
470194-720PK 30.2 USD InStock
470194-720
Prepared Eosin Methylene Blue Agar (EMB) Media
Media for Microbiology Dehydrated Media for Microbiology
This selective and differential medium is used to isolate gram negative organisms.
  • Sterile, pre-prepared media; just inoculate and incubate.
  • Selective and differential for gram negative organisms
  • Contains indicator dyes that form purple precipitate at low pH
This media contains Eosin Y and Methylene Blue, which are indicator dyes, forming a dark purple precipitate at a low pH, an indication of the presence of gram negative organisms such as fecal coliforms. This medium is not meant to act as a general purpose medium and will not support the growth of gram positive organisms.
Each bottle pours approximately 10 Petri dishes. Simply melt in a microwave oven, water bath or autoclave. Once cool enough to handle, pour into sterile Petri dish plates
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty