Would you like to log in again?
Microplates
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4-421 82.1581
470016-176PK 33.5 USD InStock
470016-176 470199-952
Microplates
Microplates HTS Microplates
These microplates replace standard test tubes and beakers. The dual-well plates are made of high-quality polystyrene and have 12 large wells and 48 small wells.
  • Closure type: slip-on lid
  • Base shape: flat base
  • Number of wells: 96
  • Max. volume: 390 µl
They are not for use with organic solvents.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty