Would you like to log in again?
Elements, Compounds, and Mixtures I Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
13-106 13-106
470200-076EA 132.5 USD InStock
470200-076 470002-476
Elements, Compounds, and Mixtures I Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Perform Two Different Techniques to Separate Mixtures.
  • Hands on separation
  • Vocabulary lesson included
Introduce students to the concepts of elements, compounds, and mixtures with special emphasis on separating mixtures by filtration and differences in solubility, chromatography, and evaporation. Students separate two mixtures by filtration and differences in solubility, one mixture using paper chromatography, and one mixture by evaporation. They isolate component colors from the chromatogram to further examine individual components. Includes all required chemicals, vocabulary exercise, reproducible worksheets and laboratory procedures. Required, but not included: balance and laboratory supplies. Designed for 24 students working in six groups of four.
Includes:
Instructor Manual
SDS
Filter Paper
Chromatography Paper
Black Ink
Copper Sulfate
Sodium Chloride
Copper Carbonate
Sulfur Powder
Teacher's Guide and Student Copymasters
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty