Would you like to log in again?
Chemical Formulas Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
13-122
470200-084CS 782.55 USD InStock
470200-084
Chemical Formulas Lab Activity
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Experimentally Determine the Correct Chemical Formulas for Various Compounds
By combining chemicals in appropriate ratios, students can determine the correct chemical formula of a new compound by observing the combination that produces the greatest amount of precipitate. Using the ratio of the combination, they can determine the chemical formula for the new substance formed. You will receive enough materials for four setups and reproducible lab and assessment manuals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty