Would you like to log in again?
Twistit® Rubber Stoppers
Supplier: PLASTICOID CO INC THE SE
G4--M299 I6--M299 J7--M299
470201-232EA 66.1 USD InStock
470201-232 470200-224 470003-668
Twistit® Rubber Stoppers
Stoppers Rubber Stoppers
An outstanding advance in rubber stoppers, four stoppers in one reduce inventory confusion and cost.
  • Twist one of the rubber protrusions off and the solid stopper becomes a one-hole stopper
  • Twist the second one off for a two-hole stopper
  • Twist the third one off for a three-hole stopper
Made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty