Would you like to log in again?
DNA Fingerprinting by Restriction Enzyme Patterns
Supplier: Edvotek
109 109
470007-368EA 153 USD InStock
470007-368 470200-476
DNA Fingerprinting by Restriction Enzyme Patterns
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
In this experiment, crime scene DNA fingerprints are compared with those obtained from suspects.
  • Required Components Included
  • Time Required: 45 minutes
  • Kit Explains Basic Concepts of DNA Fingerprinting
DNA fingerprinting is a method used in various forensic procedures as well as paternity determinations. The experiment provides the basic reagents and biologicals for DNA fragmentation pattern analysis and does not contain human DNA.
Horizontal gel electrophoresis apparatus, DC power supply, and micropipettes with tips are required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty