Would you like to log in again?
Ion Exchange Chromatography Activity Kit
Supplier: GENO TECHNOLOGY SE
BE-415
470200-750EA 207.75 USD InStock
470200-750
Ion Exchange Chromatography Activity Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Illustrate Column Chromatography Fundamentals
Most biological molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, proteins are absorbed onto a charged solid support ion-exchange column and subsequently separated during elution using a two-step salt gradient. Students learn basic principals of column chromatography and practice techniques of packing a column, eluting and collecting column fractions, and performing analysis of column fractions through spectrophotometry. Includes materials for six setups.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty