Would you like to log in again?
Laboratory Magnet Set
Supplier: United Scientific Supplies
MSET4 MSET4
470200-924EA 62.85 USD InStock
470200-924 470144-622
Laboratory Magnet Set
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A must for the Lab
  • Set of four
The magnets in this set are permanent Alnico and include two bars, two horseshoes, and keepers. Sizes: bars 3"L x 5/8"W x 3/8"H; small horseshoe 1 "W x 1"H x 3/8"D; large horseshoe 1 3/8"W x 1 3/8"H x 3/4"D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty