Would you like to log in again?
Electrostatic Demonstration Kit
Supplier: VWR International
PH0899
470201-086EA 168.15 USD InStock
470201-086
Electrostatic Demonstration Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This complete set of materials makes possible multiple dramatic demonstrations of basic electrostatic principles.
  • Experiments include operation of the electrophorus, charging the electroscope by induction and contact, attraction and repulsion, electrostatic induction, and surface distribution of charge
Includes: 2 Electroscopes, Ball and Disc Terminals for Electroscopes, Faraday Cage, Aluminum Tumbler for Ice Pail Experiment, Neon Lamp, Electrophorus with Insulating Handle, Polyethylene Film Charge Transfer Ball 15 mm, Friction Rods, Cork Ball, Pith Balls, Mounted Point for Lightning Rod Effect, acetate Cloth, acrylic Sheet, acrylic, Insulated Base for Faraday Cage, Polyvinyl Chloride, Nylon & Polystyrene, Glass.
Ordering information: Includes 2 Electroscopes, Ball and Disc Terminals for Electroscopes, Faraday Cage, Aluminum Tumbler for Ice Pail Experiment, Neon Lamp, Electrophorus with Insulating Handle, Polyethylene Film Charge Transfer Ball 15 mm, Friction Rods, Cork Ball, Pith Balls, Mounted Point for Lightning Rod Effect, acetate Cloth, acrylic Sheet, acrylic, Insulated Base for Faraday Cage, Polyvinyl Chloride, Nylon and Polystyrene, Glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty