Would you like to log in again?
In Search of My Father Kit
Supplier: Edvotek
S-49 S-49
470201-104EA 80 USD InStock
470201-104 470106-946
In Search of My Father Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
This simulation experiment demonstrates the value of mitochondrial and chromosomal DNA testing for genetic identification.
  • Activity Uses Chromosomal and Mitochondrial DNA
  • Time Requirement: 45 Minutes
  • Students Simulate a Paternity Test
  • Material for 10 Student Groups
  • Students Learn to How to Process Genetic Identification Tests
The experiment focuses on a scenario where two young boys were separated from their respective parents during a civil war. After a decade of separation, the relationship of the boys’ mothers is first determined through mitochondrial DNA testing. Additional testing of chromosomal DNA provides further information for the identification of the two boys' respective fathers.
The kit includes materials for 10 student groups to complete the activity in 45 minutes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty