Would you like to log in again?
Double Buret Clamp
Supplier: United Scientific Supplies
CLBUR2N
470201-434EA 25.5 USD InStock
470201-434
Double Buret Clamp
Clamps General Purpose Clamps
Must have for any lab.
  • Plastic covered spring jaws
  • Hold burets up to 35 mm outer diameter
  • Fits support rods up to 1/2" diameter
Overall size of this student grade, metal, Double Buret Clamp is 21 x 9.5 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty