Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PYREX® Petri Dishes with Covers
Supplier: Corning
PYREX®
3160-101 3160-60 3160-102 3160-100
470202-134CS 1016.01 USD InStock
470202-134 470202-136 470202-132 470176-008
PYREX® Petri Dishes with Covers
Culture Dishes Petri Dishes
Flat, clear dishes that can withstand repeated sterilization (wet or dry).
  • Beaded edges to provide greater mechanical strength
  • Covered dishes are not airtight
These Pyrex® Petri Dishes are perfect for repeated use in the classroom or laboratory. The edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, thereby reducing the capillary action of the condensed moisture on the sides. They are not affected chemically or thermally by any of the methods commonly employed in laboratories where sterilization is routinely used. The covered dish is not airtight. Covers are marked in blue enamel and the bottoms in white enamel to make sorting easier. Bottoms also have a triangular enamel reference point for serial dilutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty