Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Electric Timer

Supplier: DIMCOGRAY COMPANY
7-171-160R
470202-492EA 459 USD InStock
470202-492
Electric Timer
Timers Digital Timers
This sensitive lab instrument is accurate to ±0.15% at its maximum setting.

  • Maximum load: 1200 W, ¹/₃ hp
  • Range: 60 minutes (minutes/seconds)
  • Dial size: 6¹/₂"Ø

The dial’s larger numbers show elapsed time, while the smaller numbers can be used as a stop clock. It is composed of a sturdy polycarbonate case, built-in buzzer, and two separate three-wire outlet receptacles for automatic switching of appliances.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty