Would you like to log in again?
Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
Supplier: HI MEDIA LABS LTD LBS MARG
Ward®
38-1009
470202-838EA 42.75 USD InStock
470202-838
Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
Media for Microbiology Dehydrated Media for Microbiology
Dilute quantities to any volume as needed.
  • Economical alternative
  • Stores much longer than prepared media
  • Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty