Would you like to log in again?
LaMotte® Carbon Dioxide Water Test Kit
Supplier: LaMotte
7297-DR-01 R-7297-DR
470203-124EA 65.2 USD InStock
470203-124 470021-200
LaMotte® Carbon Dioxide Water Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range: 0 to 50 ppm
  • Materials for 50 tests
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty