Would you like to log in again?
Ward's® DataHub Mobile Charging Cart
Ward's
LD-CART LD-CART
470225-188EA 1599.99 USD InStock
470225-188 470210-542
Ward's® DataHub Mobile Charging Cart
Store, move and charge your Datahubs.
  • Holds 16 Datahubs
  • 16 slots for Ipads or Tablets
  • Easy to roll from class to class
This unique storage environment for Ward's DataHub products contains everything needed for STEM-based hands-on science activities, all in a single, clean and organized science station on wheels. Built-in charger 5 V and 6 V output. Total power 350 W. Power cord is 110/220 V, comes with a US plug. Size: 691×526×526 mm. Weight empty: 27 kg.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty