Would you like to log in again?
Simple Machines, Inclined Plane/Cart Model
Supplier: EISCO SCIENTIFIC LLC
WDMS10
470210-806EA 45.45 USD InStock
470210-806
Simple Machines, Inclined Plane/Cart Model
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This wooden machine is elegantly designed and easy for students to use.
  • Cart with built-in mass
  • Additional mass can be added
  • Includes manual
It comes with a built-in mass, but the cart has a shallow indent for adding additional mass if needed. Students can study several different simple machine concepts with this one apparatus. Includes an instruction manual to facilitate student and teacher use of the apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty