Would you like to log in again?
Simple Machines, Simple Wedge Model
Supplier: EISCO SCIENTIFIC LLC
WDMS14
470210-814EA 44.4 USD InStock
470210-814
Simple Machines, Simple Wedge Model
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This wooden machine is elegantly designed to demonstrate how a wedge can increase mechanical advantage.
  • Demonstrate mechanical advantages
  • Includes manual
Easy for students to use and measure. Includes an instruction manual to facilitate student and teacher use of the apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty