Would you like to log in again?
Simple Machines, Fulcrum Balance Model
Supplier: EISCO SCIENTIFIC LLC
WDMS17
470210-820EA 35.4 USD InStock
470210-820
Simple Machines, Fulcrum Balance Model
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This wooden machine is elegantly designed and easy for students to use. Study how changing the distance from the fulcrum changes the mechanical advantage.
Students can intuitively figure out the balance point for a given set of masses. Includes an instruction manual to facilitate student and teacher use of the apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty