Would you like to log in again?
Simple Machines, Screw Model
Supplier: EISCO SCIENTIFIC LLC
WDMS20
470210-826EA 47.6 USD InStock
470210-826
Simple Machines, Screw Model
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This wooden machine has a sturdy wooden construction ideal for classroom use.
  • Demonstrate the concept of leverage
  • Includes manual
The large block mounted around the head of the screw gives students additional leverage so they can easily break eggs or squish other objects with the screw that would be difficult to break with their bare hands. The square top also makes it easy to measure the distance top of the screw is turned vs. the distance the screw is sent into the board. Includes an instruction manual to facilitate students and teacher use of the apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty