Would you like to log in again?
Eisco® Median Section of the Head
Supplier: EISCO SCIENTIFIC LLC
AM18AS
470212-446EA 218.25 USD InStock
470212-446
Eisco® Median Section of the Head
Models Human Biology Models
A highly detailed model that identifies 70 structures in a typical human head and neck.
  • Life size, median section of a human head and neck
  • Hand painted
  • Flat mounted on base
  • Includes numbered key card
Presented as a sagittal plane section of the anatomy within the human head. This full size model allows you to easily examine the anatomy of the brain, nose, mouth, pharynx, larynx, vertebral column, and neck.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty