Would you like to log in again?
Eisco® Giant Heart Model
Supplier: EISCO SCIENTIFIC LLC
AM75AS
470212-476EA 279.35 USD InStock
470212-476
Eisco® Giant Heart Model
Models Human Biology Models
Get an amazing look at the human heart with this 3 times life size model. Dissectible into 7 parts, it shows excellent detail of both internal and external structures. Great for study of the human circulation and the movement of blood through the heart. 
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty