Would you like to log in again?
Visual Scientifics Atwood Machine
Supplier: EISCO SCIENTIFIC LLC
PTAWM
470212-490EA 36.65 USD InStock
470212-490
Visual Scientifics Atwood Machine
Educational Materials Physics Educational Materials Mechanics Learning Activities
Take your study of pulleys and engineering to the next level!
  • Help students dispell misconceptions about tension and Newton's laws
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
  • 10+ page instruction book included
Once students have an understanding of Newton’s laws and understand tension and using pulley’s to change direction of force this module is the next level of merging engineering and inquiry together. It is often difficult for students to conceptualize exactly how pulleys work. One common misconception which causes problems when students try to construct free body diagram and solve problems using pulleys is the idea of tension in a string. In this modules students will work though activities to solve this misconception, such as placing a spring between 2 pulleys. Also comes with a 10+ page instruction booklet covering approximately 2 to 3 hours of class/lab time material. Instructions are written to explicitly cover Next Generation Science Standards Concepts, and Common Core Standards. Every module includes comprehensive instructions to assist with lesson planning, student activities, and experimental procedures. All labs and extensions feature recommended video analysis setups and guides. Included items are Atwood machine, right angle clamp, string, spring, support post. Required but not included 2.5 N spring scale or force probe, Hooked mass set (100 g) (9 to 10 g weights and 1 to 10 g hook), Video recording device, Visual Scientifics base and Visual Scientifics backboard.
Ordering information: Visual Scientifics Base (470148-822) and Backboard (470135-558) required, but sold separately. Items included in this kit: Atwood machine, Right angle clamp, String, Spring, and Support post. Required but not included: 2.5 N spring scale or force probe, 100 g Hooked mass set (9 to 10 gram weights and 1 to 10 g hook), Video recording device, Visual Scientifics base, and Visual Scientifics Back Board.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty